missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxy-4-methylphenol, 97%

Catalog No. AAB2180114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2180114 25 g
AAB2180122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2180114 Supplier Thermo Scientific Chemicals Supplier No. B2180114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6638-05-7 | C9H12O3 | 168.19 g/mol

Used as a flavor and fragrance agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavor and fragrance agents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 6638-05-7
MDL Number MFCD00017289

Specifications

Melting Point 36°C to 42°C
Boiling Point 144°C to 146°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Phenol-like
Molecular Formula C16H13NO3
Quantity 25 g
Synonym 5-benzyloxy-1h-indole-2-carboxylic acid, 5-benzyloxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-phenylmethoxy, chembl24850, 5-phenylmethoxy-1h-indole-2-carboxylic acid, 5-phenylmethoxy indole-2-carboxylic acid, 5-benzyloxy-1h-indole-2-carboxylicacid, maybridge1_005333, acmc-209x2s
Solubility Information Slightly soluble in water.
InChI Key MVCLSAMNMAWXFQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COC2=CC3=C(C=C2)NC(=C3)C(=O)O
IUPAC Name 5-phenylmethoxy-1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 267.28
PubChem CID 81146
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 2,6-Dimethoxy-4-methylphenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-641-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only