missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dihydroxyacetophenone 97.0+%, TCI America™

Catalog No. p-7126893
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17165G 5 g
D171625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17165G Supplier TCI America Supplier No. D17165G
Only null left

Chemical Identifiers

CAS 699-83-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002270
InChI Key YPTJKHVBDCRKNF-UHFFFAOYSA-N
Synonym 2',6'-dihydroxyacetophenone, 1-2,6-dihydroxyphenyl ethanone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, resorcinol, 2-acetyl, 1-2,6-dihydroxyphenyl ethan-1-one, 2,6-dihydroxy acetophenone, gamma-resacetophenone, ethanone, 1-2,6-dihydroxyphenyl, unii-88bo51g3y2
PubChem CID 69687
IUPAC Name 1-(2,6-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC=C1O)O

Specifications

Melting Point 155°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,6′-Dihydroxyacetophenone
EINECSNumber (3)-1250
TSCA No