missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dihydroxyacetophenone, 98+%

CAS: 699-83-2 | C8H8O3 | 152.149 g/mol

Supplier:  Thermo Scientific Chemicals A1462714

 View more versions of this product

Catalog No. AAA1462714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2',6'-Dihydroxyacetophenone
156°C to 159°C
MFCD00002270
1366061
YPTJKHVBDCRKNF-UHFFFAOYSA-N
1-(2,6-dihydroxyphenyl)ethanone
69687
≥98%
699-83-2
C8H8O3
25 g
2',6'-dihydroxyacetophenone, 1-2,6-dihydroxyphenyl ethanone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, resorcinol, 2-acetyl, 1-2,6-dihydroxyphenyl ethan-1-one, 2,6-dihydroxy acetophenone, gamma-resacetophenone, ethanone, 1-2,6-dihydroxyphenyl, unii-88bo51g3y2
CC(=O)C1=C(C=CC=C1O)O
152.149
152.15

Chemical Identifiers

699-83-2
152.149
YPTJKHVBDCRKNF-UHFFFAOYSA-N
69687
CC(=O)C1=C(C=CC=C1O)O
C8H8O3
MFCD00002270
2',6'-dihydroxyacetophenone, 1-2,6-dihydroxyphenyl ethanone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, resorcinol, 2-acetyl, 1-2,6-dihydroxyphenyl ethan-1-one, 2,6-dihydroxy acetophenone, gamma-resacetophenone, ethanone, 1-2,6-dihydroxyphenyl, unii-88bo51g3y2
1-(2,6-dihydroxyphenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-833-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only