missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dihydroxyacetophenone, 98+%

Catalog No. AAA1462722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1462722 100 g
AAA1462706 5 g
AAA1462714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1462722 Supplier Thermo Scientific Chemicals Supplier No. A1462722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 699-83-2 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 699-83-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002270
InChI Key YPTJKHVBDCRKNF-UHFFFAOYSA-N
Synonym 2',6'-dihydroxyacetophenone, 1-2,6-dihydroxyphenyl ethanone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, resorcinol, 2-acetyl, 1-2,6-dihydroxyphenyl ethan-1-one, 2,6-dihydroxy acetophenone, gamma-resacetophenone, ethanone, 1-2,6-dihydroxyphenyl, unii-88bo51g3y2
PubChem CID 69687
IUPAC Name 1-(2,6-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC=C1O)O

Specifications

Melting Point 156°C to 159°C
Quantity 100 g
Beilstein 1366061
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2',6'-Dihydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-833-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only