missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Difluoropyridine 98.0+%, TCI America™
$65.46 - $182.89
Chemical Identifiers
| CAS | 1513-65-1 |
|---|---|
| Molecular Formula | C5H3F2N |
| Molecular Weight (g/mol) | 115.083 |
| MDL Number | MFCD00006227 |
| InChI Key | MBTGBRYMJKYYOE-UHFFFAOYSA-N |
| Synonym | pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine |
| PubChem CID | 73934 |
| IUPAC Name | 2,6-difluoropyridine |
| SMILES | C1=CC(=NC(=C1)F)F |
Chemical Identifiers
| 1513-65-1 | |
| 115.083 | |
| MBTGBRYMJKYYOE-UHFFFAOYSA-N | |
| 73934 | |
| C1=CC(=NC(=C1)F)F |
| C5H3F2N | |
| MFCD00006227 | |
| pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine | |
| 2,6-difluoropyridine |
Specifications
| 1513-65-1 | |
| 125°C | |
| MFCD00006227 | |
| 1993 | |
| MBTGBRYMJKYYOE-UHFFFAOYSA-N | |
| 2,6-difluoropyridine | |
| 73934 | |
| ≥98.0% (GC) | |
| 2,6-Difluoropyridine |
| Yellow | |
| C5H3F2N | |
| 5 g | |
| pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine | |
| C1=CC(=NC(=C1)F)F | |
| 115.083 | |
| 115.08 | |
| Liquid |
Safety and Handling
TSCA : No