missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoropyridine, 99%

Catalog No. AC306660050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC306660050 5 g Glass bottle
AC306660250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC306660050 Supplier Thermo Scientific Chemicals Supplier No. 306660050

CAS: 1513-65-1 | C5H3F2N | 115.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1513-65-1
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.08
MDL Number MFCD00006227
InChI Key MBTGBRYMJKYYOE-UHFFFAOYSA-N
Synonym pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine
PubChem CID 73934
IUPAC Name 2,6-difluoropyridine
SMILES C1=CC(=NC(=C1)F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Purple-Red
Density 1.2600g/mL
Boiling Point 124.5°C (743.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380
Quantity 5 g
Specific Gravity 1.26
Formula Weight 115.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Difluoropyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If eye irritation persists: Get medical advice/a
EINECSNumber 216-152-8

RUO – Research Use Only