Learn More
2,6-Difluoropyridine, 98+%
CAS: 1513-65-1 | C5H3F2N | 115.083 g/mol
$73.97 - $933.85
Chemical Identifiers
| CAS | 1513-65-1 |
|---|---|
| Molecular Formula | C5H3F2N |
| Molecular Weight (g/mol) | 115.083 |
| MDL Number | MFCD00006227 |
| InChI Key | MBTGBRYMJKYYOE-UHFFFAOYSA-N |
| Synonym | pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine |
| PubChem CID | 73934 |
| IUPAC Name | 2,6-difluoropyridine |
| SMILES | C1=CC(=NC(=C1)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1552206
|
Thermo Scientific Chemicals
A1552206 |
5 g |
Each for $73.97
|
|
|||||
|
AAA1552214
|
Thermo Scientific Chemicals
A1552214 |
25 g |
Each for $285.99
|
|
|||||
|
AAA1552222
|
Thermo Scientific Chemicals
A1552222 |
100 g |
Each for $933.85
|
|
|||||
Description
2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.
Solubility
Not miscible or difficult to mix.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Chemical Identifiers
| 1513-65-1 | |
| 115.083 | |
| MBTGBRYMJKYYOE-UHFFFAOYSA-N | |
| 73934 | |
| C1=CC(=NC(=C1)F)F |
| C5H3F2N | |
| MFCD00006227 | |
| pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine | |
| 2,6-difluoropyridine |
Specifications
| 1513-65-1 | |
| 123°C to 124°C | |
| Characteristic | |
| 1.437 | |
| 5 g | |
| 1422549 | |
| Not miscible or difficult to mix. | |
| C1=CC(=NC(=C1)F)F | |
| 115.083 | |
| 115.08 | |
| 2,6-Difluoropyridine |
| 1.268 | |
| 33°C (91°F) | |
| C5H3F2N | |
| MFCD00006227 | |
| UN1993 | |
| pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine | |
| MBTGBRYMJKYYOE-UHFFFAOYSA-N | |
| 2,6-difluoropyridine | |
| 73934 | |
| ≥98% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 216-152-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only