missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoropyridine, 98+%

Catalog No. AAA1552222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1552222 100 g
AAA1552206 5 g
AAA1552214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1552222 Supplier Thermo Scientific Chemicals Supplier No. A1552222

CAS: 1513-65-1 | C5H3F2N | 115.083 g/mol

2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.

Solubility
Not miscible or difficult to mix.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 1513-65-1
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD00006227
InChI Key MBTGBRYMJKYYOE-UHFFFAOYSA-N
Synonym pyridine, 2,6-difluoro, 2,6-difluorpyridin, ccris 1720, 2,6-difluoro-pyridine, 2,6-difluoropyridin, pubchem9241, 2,6 difluoropyridine, 2, 6-difluoropyridine, 2,6 difluoro pyridine, 2,6-di-fluoropyridine
PubChem CID 73934
IUPAC Name 2,6-difluoropyridine
SMILES C1=CC(=NC(=C1)F)F

Specifications

Density 1.268
Boiling Point 123°C to 124°C
Flash Point 33°C (91°F)
Odor Characteristic
Refractive Index 1.437
Quantity 100 g
UN Number UN1993
Beilstein 1422549
Solubility Information Not miscible or difficult to mix.
Formula Weight 115.08
Percent Purity ≥98%
Chemical Name or Material 2,6-Difluoropyridine
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 216-152-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only