missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. D25765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25765G Supplier TCI America Supplier No. D25765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85118-00-9
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD00000329
InChI Key LSXJPJGBWSZHTM-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl bromide, 2-bromomethyl-1,3-difluorobenzene, 2,6-difluorobenzylbromide, alpha-bromo-2,6-difluorotoluene, benzene, 2-bromomethyl-1,3-difluoro, timtec-bb sbb006564, 2-bromomethyl-1,3-difluoro-benzene, fr cf b1e, pubchem4915, acmc-209q4j
PubChem CID 581435
IUPAC Name 2-(bromomethyl)-1,3-difluorobenzene
SMILES FC1=CC=CC(F)=C1CBr

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 207.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluorobenzyl Bromide