missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. D29405G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29405G 5 g
D29401G 1 g
D294025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D29405G Supplier TCI America Supplier No. D29405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60230-36-6
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00085004
InChI Key QXWAUQMMMIMLTO-UHFFFAOYSA-N
Synonym 2,6-difluorobenzene-1-sulfonyl chloride, 2,6-difluorobenzenesulfonylchloride, 2,6-difluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,6-difluoro, 2,6-difluoro-benzenesulfonyl chloride, 2,6-difluorobenzene sulfonyl chloride, 2,6-difluorophenyl chlorosulfone, pubchem1957, acmc-209mil, ksc352q0t
PubChem CID 2734271
IUPAC Name 2,6-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)F)S(=O)(=O)Cl)F

Specifications

Color Yellow
Boiling Point 129°C
Quantity 5 g
UN Number 3265
Formula Weight 212.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzenesulfonyl Chloride