missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',6'-Difluoroacetophenone 98.0+%, TCI America™
$121.16 - $486.52
Chemical Identifiers
| CAS | 13670-99-0 |
|---|---|
| Molecular Formula | C8H6F2O |
| Molecular Weight (g/mol) | 156.13 |
| MDL Number | MFCD00000328 |
| InChI Key | VGIIILXIQLXVLC-UHFFFAOYSA-N |
| Synonym | 2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054 |
| PubChem CID | 83643 |
| IUPAC Name | 1-(2,6-difluorophenyl)ethan-1-one |
| SMILES | CC(=O)C1=C(F)C=CC=C1F |
Chemical Identifiers
| 13670-99-0 | |
| 156.13 | |
| VGIIILXIQLXVLC-UHFFFAOYSA-N | |
| 83643 | |
| CC(=O)C1=C(F)C=CC=C1F |
| C8H6F2O | |
| MFCD00000328 | |
| 2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054 | |
| 1-(2,6-difluorophenyl)ethan-1-one |
Specifications
| 13670-99-0 | |
| 79°C | |
| MFCD00000328 | |
| 2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054 | |
| CC(=O)C1=C(F)C=CC=C1F | |
| 156.13 | |
| 156.13 | |
| Liquid |
| Yellow | |
| C8H6F2O | |
| 5 g | |
| VGIIILXIQLXVLC-UHFFFAOYSA-N | |
| 1-(2,6-difluorophenyl)ethan-1-one | |
| 83643 | |
| ≥98.0% (GC) | |
| 2′,6′-Difluoroacetophenone |
Safety and Handling
TSCA : No