missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Difluoroacetophenone 98.0+%, TCI America™

Catalog No. D35595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35595G 5 g
D355925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35595G Supplier TCI America Supplier No. D35595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13670-99-0
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00000328
InChI Key VGIIILXIQLXVLC-UHFFFAOYSA-N
Synonym 2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
PubChem CID 83643
IUPAC Name 1-(2,6-difluorophenyl)ethan-1-one
SMILES CC(=O)C1=C(F)C=CC=C1F

Specifications

Color Yellow
Boiling Point 79°C
Quantity 5 g
Formula Weight 156.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,6′-Difluoroacetophenone