missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diethynylpyridine 98.0+%, TCI America™

Catalog No. D5097200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5097200MG 200 mg
D50971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5097200MG Supplier TCI America Supplier No. D5097200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75867-46-8
Molecular Formula C9H5N
Molecular Weight (g/mol) 127.146
MDL Number MFCD09039127
InChI Key VYRLFYTZNXGQIK-UHFFFAOYSA-N
PubChem CID 12579661
IUPAC Name 2,6-diethynylpyridine
SMILES C#CC1=NC(=CC=C1)C#C

Specifications

Melting Point 71°C
Quantity 200 mg
Formula Weight 127.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diethynylpyridine
TSCA No
Recommended Storage Refrigerator