missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diethylphenyl isocyanate, 98+%

Catalog No. aal1133906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1133906 5 g
AAL1133903 1 g
AAL1133914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AAL1133906 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' L1133906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20458-99-5 | C11H13NO | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20458-99-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00019912
InChI Key AUSRFKVPUCQCCN-UHFFFAOYSA-N
Synonym 2,6-diethylphenyl isocyanate, 2,6-diethylphenylisocyanate, 1,3-diethyl-2-isocyanato-benzene, benzene, 1,3-diethyl-2-isocyanato, acmc-1co7s, 2,6-diethylbenzenisocyanate, benzene,1,3-diethyl-2-isocyanato
PubChem CID 5153183
IUPAC Name 1,3-diethyl-2-isocyanatobenzene
SMILES CCC1=CC=CC(CC)=C1N=C=O

Specifications

Density 1.014
Boiling Point 114°C to 115°C (1 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.526
Quantity 5 g
UN Number UN2206
Beilstein 2641462
Sensitivity Moisture sensitive
Formula Weight 175.23
Percent Purity ≥98%
Chemical Name or Material 2,6-Diethylphenyl isocyanate
Show More Show Less
missing translation for 'healthHazard1' H301-H315-H319-H330-H334-H335
missing translation for 'healthHazard2' GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P260-P264b-P270-P271-P280-P284-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only