missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloropyridine 97.0+%, TCI America™
$29.83 - $173.37
Chemical Identifiers
| CAS | 2402-78-0 |
|---|---|
| Molecular Formula | C5H3Cl2N |
| Molecular Weight (g/mol) | 147.99 |
| MDL Number | MFCD00006244 |
| InChI Key | FILKGCRCWDMBKA-UHFFFAOYSA-N |
| Synonym | pyridine, 2,6-dichloro, ccris 1727, 2,6-dichlorpyridine, 2.6-dichloropyridine, pubchem2564, 2,6-dichloro pyridine, 2,6-dichloro-pyridine, acmc-1cbwr, 26dclpy, wln: t6nj bg fg |
| PubChem CID | 16989 |
| IUPAC Name | 2,6-dichloropyridine |
| SMILES | ClC1=CC=CC(Cl)=N1 |
Chemical Identifiers
| 2402-78-0 | |
| 147.99 | |
| FILKGCRCWDMBKA-UHFFFAOYSA-N | |
| 16989 | |
| ClC1=CC=CC(Cl)=N1 |
| C5H3Cl2N | |
| MFCD00006244 | |
| pyridine, 2,6-dichloro, ccris 1727, 2,6-dichlorpyridine, 2.6-dichloropyridine, pubchem2564, 2,6-dichloro pyridine, 2,6-dichloro-pyridine, acmc-1cbwr, 26dclpy, wln: t6nj bg fg | |
| 2,6-dichloropyridine |
Specifications
| 2402-78-0 | |
| White-Yellow | |
| C5H3Cl2N | |
| 25 g | |
| pyridine, 2,6-dichloro, ccris 1727, 2,6-dichlorpyridine, 2.6-dichloropyridine, pubchem2564, 2,6-dichloro pyridine, 2,6-dichloro-pyridine, acmc-1cbwr, 26dclpy, wln: t6nj bg fg | |
| ClC1=CC=CC(Cl)=N1 | |
| 147.99 | |
| 147.99 | |
| Crystalline Powder |
| 86°C | |
| 212°C | |
| MFCD00006244 | |
| 2811 | |
| FILKGCRCWDMBKA-UHFFFAOYSA-N | |
| 2,6-dichloropyridine | |
| 16989 | |
| ≥97.0% (GC) | |
| 2,6-Dichloropyridine |
Safety and Handling
EINECSNumber : (5)-3688
RTECSNumber : US8400000
TSCA : Yes