missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine 97.0+%, TCI America™

Catalog No. D041025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D041025G 25 g
D0410500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D041025G Supplier TCI America Supplier No. D041025G

Chemical Identifiers

CAS 2402-78-0
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006244
InChI Key FILKGCRCWDMBKA-UHFFFAOYSA-N
Synonym pyridine, 2,6-dichloro, ccris 1727, 2,6-dichlorpyridine, 2.6-dichloropyridine, pubchem2564, 2,6-dichloro pyridine, 2,6-dichloro-pyridine, acmc-1cbwr, 26dclpy, wln: t6nj bg fg
PubChem CID 16989
IUPAC Name 2,6-dichloropyridine
SMILES ClC1=CC=CC(Cl)=N1

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 212°C
Quantity 25 g
UN Number 2811
Formula Weight 147.99
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloropyridine
EINECSNumber (5)-3688
RTECSNumber US8400000
TSCA Yes