missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyrazine 99.0+%, TCI America™

Catalog No. p-7126978
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
D18675G 5 g
D186725G 25 g
2 options

Catalog No. D18675G

Supplier: TCI America D18675G

Only null left

Chemical Identifiers

CAS 4774-14-5
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00006125
InChI Key LSEAAPGIZCDEEH-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dichloro, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazine, 2-chloro-6-chloro-pyrazine, 2,6-dicloropyrazine, 2,6 dichloropyrazine, pubchem8548, 2,6-dichloro-pyrazin, 2, 6-dichloropyrazine
PubChem CID 78504
IUPAC Name 2,6-dichloropyrazine
SMILES C1=C(N=C(C=N1)Cl)Cl

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 148.97
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloropyrazine