missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$73.16 - $73.16
Chemical Identifiers
| CAS | 73852-17-2 |
|---|---|
| Molecular Formula | C6H5BCl2O2 |
| Molecular Weight (g/mol) | 190.81 |
| MDL Number | MFCD00064869 |
| InChI Key | CXDPUSMFYPQXCV-UHFFFAOYSA-N |
| PubChem CID | 2734332 |
| IUPAC Name | (2,6-dichlorophenyl)boronic acid |
| SMILES | OB(O)C1=C(Cl)C=CC=C1Cl |
Chemical Identifiers
| 73852-17-2 | |
| 190.81 | |
| CXDPUSMFYPQXCV-UHFFFAOYSA-N | |
| (2,6-dichlorophenyl)boronic acid |
| C6H5BCl2O2 | |
| MFCD00064869 | |
| 2734332 | |
| OB(O)C1=C(Cl)C=CC=C1Cl |
Specifications
| 73852-17-2 | |
| White-Yellow | |
| MFCD00064869 | |
| 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid | |
| OB(O)C1=C(Cl)C=CC=C1Cl | |
| 190.81 | |
| 190.81 | |
| 2,6-Dichlorophenylboronic Acid (contains varying amounts of Anhydride) |
| 151°C | |
| C6H5BCl2O2 | |
| 1 g | |
| CXDPUSMFYPQXCV-UHFFFAOYSA-N | |
| (2,6-dichlorophenyl)boronic acid | |
| 2734332 | |
| Crystalline Powder |
Safety and Handling
TSCA : No