missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenethyl alcohol, 96%

Catalog No. p-106370
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350760050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC350760050 Supplier Thermo Scientific Chemicals Supplier No. 350760050

CAS: 30595-79-0 | C8H8Cl2O | 191.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30595-79-0
Molecular Formula C8H8Cl2O
Molecular Weight (g/mol) 191.05
MDL Number MFCD00800674
InChI Key ZBQPKQUIKJDGIX-UHFFFAOYSA-N
Synonym 2,6-dichlorophenethyl alcohol, 2-2,6-dichlorophenyl ethanol, 2,6-dichlorophenethylalcohol, 2-2,6-dichlorophenyl ethan-1-ol, 2,6-dichlorophenethanol, benzeneethanol, 2,6-dichloro, acmc-209hha, 2,6-dichlorobenzeneethanol, ksc493m0h, benzeneethanol,2,6-dichloro
PubChem CID 2734100
IUPAC Name 2-(2,6-dichlorophenyl)ethanol
SMILES OCCC1=C(Cl)C=CC=C1Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 59°C to 65°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 191.06
Percent Purity 96%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2, 6-Dichlorophenethyl alcohol, 96%
Show More Show Less

RUO – Research Use Only