missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloronicotinic Acid 98.0+%, TCI America™

Catalog No. D34125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34125G 5 g
D341225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34125G Supplier TCI America Supplier No. D34125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38496-18-3 | C6H3Cl2NO2 | 191.995 g/mol

Chemical Identifiers

CAS 38496-18-3
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00075583
InChI Key AJPKQSSFYHPYMH-UHFFFAOYSA-N
Synonym 2,6-dichloronicotinic acid, 2,6-dichloro-nicotinic acid, 2,6-dichloro-3-pyridinecarboxylic acid, dichloronicotinic acid, 2,6-dichloronicotininc acid, 3-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloronicotinincacid, pubchem1225, 2,6-dichlornicotinsaure, acmc-209j0g
PubChem CID 854047
IUPAC Name 2,6-dichloropyridine-3-carboxylic acid
SMILES C1=CC(=NC(=C1C(=O)O)Cl)Cl

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Formula Weight 192.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloronicotinic Acid