missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloroisonicotinic Acid 98.0+%, TCI America™
$55.84 - $157.87
Chemical Identifiers
| CAS | 5398-44-7 |
|---|---|
| Molecular Formula | C6H3Cl2NO2 |
| Molecular Weight (g/mol) | 192.00 |
| MDL Number | MFCD00234147 |
| InChI Key | SQSYNRCXIZHKAI-UHFFFAOYSA-N |
| Synonym | 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403 |
| PubChem CID | 94830 |
| ChEBI | CHEBI:73179 |
| IUPAC Name | 2,6-dichloropyridine-4-carboxylic acid |
| SMILES | OC(=O)C1=CC(Cl)=NC(Cl)=C1 |
Chemical Identifiers
| 5398-44-7 | |
| 192.00 | |
| SQSYNRCXIZHKAI-UHFFFAOYSA-N | |
| 94830 | |
| 2,6-dichloropyridine-4-carboxylic acid |
| C6H3Cl2NO2 | |
| MFCD00234147 | |
| 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403 | |
| CHEBI:73179 | |
| OC(=O)C1=CC(Cl)=NC(Cl)=C1 |
Specifications
| 5398-44-7 | |
| C6H3Cl2NO2 | |
| 1 g | |
| SQSYNRCXIZHKAI-UHFFFAOYSA-N | |
| 2,6-dichloropyridine-4-carboxylic acid | |
| 94830 | |
| 192.00 | |
| Crystalline Powder |
| 211°C | |
| MFCD00234147 | |
| 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403 | |
| OC(=O)C1=CC(Cl)=NC(Cl)=C1 | |
| 192.00 | |
| CHEBI:73179 | |
| ≥98.0% (GC,T) | |
| 2,6-Dichloroisonicotinic Acid |
Safety and Handling
TSCA : No