missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol Solution, 0.001 Molar, Volumetric, Reagents

Catalog No. 01335423
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335423 1 L
01335422 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335423 Supplier Labchem Inc Supplier No. CD2601001A

2,6-Dichloroindophenol Solution, 0.001 Molar, Volumetric, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material 2,6-Dichloroindophenol Solution, 0.001 Molar, Volumetric
CAS 7732-18-5, 620-45-1
Color Deep Blue
Boiling Point 100°C
Packaging Poly Bottle
Quantity 1 L
Specific Gravity 1
Grade Reagent
Physical Form Liquid