missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol sodium salt hydrate, Honeywell Fluka™

Catalog No. p-7072190
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
6002650 5 g
1 options

Catalog No. 6002650

Supplier: Honeywell Chemicals 361805G

Only null left

≥97.0% (calc. based on dry substance, AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Quantity 5 g
UN Number NONH for all modes of transport
Beilstein 3641229
Formula Weight 290.08g/mol (anhydrous basis)
Percent Purity ≥97% (calculated based on dry substance, AT)
Chemical Name or Material 2,6-Dichloroindophenol sodium salt hydrate
RTECSNumber GU5495000