missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloroindophenol, sodium salt hydrate, 90+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Assay Percent Range | 90+% |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Solubility Information | Solubility in water: soluble. Other solubilities: freely soluble in alcohol |
| Formula Weight | 290.07 |
| Percent Purity | 90+% |
| Chemical Name or Material | 2, 6-Dichloroindophenol, sodium salt hydrate |
RUO – Research Use Only