missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloroindophenol Sodium Salt Dihydrate 95.0+%, TCI America™
Supplier: TCI America D037525G
Specifications
| 2,6-Dichloroindophenol Sodium Salt Dihydrate | |
| Green | |
| MFCD00150014 | |
| 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt | |
| [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 | |
| 290.07 | |
| CHEBI:948 | |
| ≥95.0% (T) |
| 620-45-1 | |
| C12H6Cl2NNaO2 | |
| 25 g | |
| FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
| sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate | |
| 23697355 | |
| 290.07 | |
| Crystalline Powder |
Chemical Identifiers
| 620-45-1 | |
| 290.07 | |
| FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
| 23697355 | |
| sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate |
| C12H6Cl2NNaO2 | |
| MFCD00150014 | |
| 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt | |
| CHEBI:948 | |
| [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 |
Safety and Handling
RTECSNumber : GU5495000
TSCA : Yes