missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol Sodium Salt, ACS, Spectrum™ Chemical

Catalog No. p-7165452
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Amber Glass Bottle
Catalog No. Quantity Packaging
18608171 1 g Amber Glass Bottle
1 options

Catalog No. 18608171

Supplier: Spectrum Chemical Mfg Cor D10221GM

Only null left

Chemical Identifiers

CAS 620-45-1

Specifications

CAS Max % 1
Loss on Drying 0.12
Packaging Amber Glass Bottle
Quantity 1 g
Grade ACS

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.