missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzothiazole 98.0+%, TCI America™

Catalog No. D38015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38015G 5 g
D38011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38015G Supplier TCI America Supplier No. D38015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3622-23-9
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.068
MDL Number MFCD00044101
InChI Key QDZGJGWDGLHVNK-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzothiazole, 2,6-dichlorobenzo d thiazole, benzothiazole, 2,6-dichloro, 2,6-dichloro-benzothiazole, 2,6-dichloro benzothiazole, pubchem17324, acmc-1adss, ksc490o9j
PubChem CID 77176
IUPAC Name 2,6-dichloro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Cl)SC(=N2)Cl

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 204.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzothiazole