missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzothiazole, 97%

Catalog No. AAL1025603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1025603 1 g
AAL1025606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1025603 Supplier Thermo Scientific Chemicals Supplier No. L1025603

CAS: 3622-23-9 | C7H3Cl2NS | 204.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3622-23-9
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.068
MDL Number MFCD00044101
InChI Key QDZGJGWDGLHVNK-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzothiazole, 2,6-dichlorobenzo d thiazole, benzothiazole, 2,6-dichloro, 2,6-dichloro-benzothiazole, 2,6-dichloro benzothiazole, pubchem17324, acmc-1adss, ksc490o9j
PubChem CID 77176
IUPAC Name 2,6-dichloro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Cl)SC(=N2)Cl

Specifications

Melting Point 94°C to 98°C
Quantity 1 g
Beilstein 127749
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorobenzothiazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-819-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only