missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzenesulfonyl Chloride 97.0+%, TCI America™

Catalog No. D42225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42225G 5 g
D42221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42225G Supplier TCI America Supplier No. D42225G
Only null left

Chemical Identifiers

CAS 6579-54-0
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00052311
InChI Key WGGKQIKICKLWGN-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzene-1-sulfonyl chloride, 2,6-dichlorobenzenesulfonylchloride, 2,6-dichlorophenylsulfonyl chloride, 2,6-dichlorobenzenesulphonyl chloride, unii-8523kf52zn, benzenesulfonyl chloride, 2,6-dichloro, 2,6-dichloro-benzenesulfonyl chloride, 2,6-dichlorobenzene sulfonyl chloride, pubchem5117, acmc-209ns8
PubChem CID 522183
IUPAC Name 2,6-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)S(=O)(=O)Cl)Cl

Specifications

Melting Point 54°C
Color Yellow
Quantity 5 g
UN Number 3261
Formula Weight 245.50
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzenesulfonyl Chloride