Learn More
2,6-Dichlorobenzeneboronic acid, 95%, Thermo Scientific Chemicals
$131.15 - $131.15
Chemical Identifiers
| CAS | 73852-17-2 |
|---|---|
| Molecular Formula | C6H5BCl2O2 |
| Molecular Weight (g/mol) | 190.81 |
| MDL Number | MFCD00064869 |
| InChI Key | CXDPUSMFYPQXCV-UHFFFAOYSA-N |
| Synonym | 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid |
| PubChem CID | 2734332 |
| IUPAC Name | (2,6-dichlorophenyl)boronic acid |
| SMILES | OB(O)C1=C(Cl)C=CC=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1981603
|
Thermo Scientific Chemicals
L1981603 |
1 g |
Each for $131.15
|
|
|||||
Description
Reagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReagent used for palladium-catalyzed stille and suzuki-miyaura cross-couplings, palladium (pd)-catalytized reactions. Reagent used in preparation of common building blocks for pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 73852-17-2 | |
| 190.81 | |
| CXDPUSMFYPQXCV-UHFFFAOYSA-N | |
| 2734332 | |
| OB(O)C1=C(Cl)C=CC=C1Cl |
| C6H5BCl2O2 | |
| MFCD00064869 | |
| 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid | |
| (2,6-dichlorophenyl)boronic acid |
Specifications
| 73852-17-2 | |
| C6H5BCl2O2 | |
| 1 g | |
| 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, 2,6-dichlorophenyl boranediol, boronic acid, 2,6-dichlorophenyl, pubchem7938, acmc-1bc3m, ksc489k8l, 2,6-dichloro-phenylboronic acid | |
| CXDPUSMFYPQXCV-UHFFFAOYSA-N | |
| (2,6-dichlorophenyl)boronic acid | |
| 2734332 | |
| 95% |
| 151°C to 152°C | |
| MFCD00064869 | |
| 4802506 | |
| Insoluble in water. | |
| OB(O)C1=C(Cl)C=CC=C1Cl | |
| 190.81 | |
| 190.82 | |
| 2,6-Dichlorobenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only