missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzaldoxime, 97%

Catalog No. p-7081399
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1135106 5 g
AAA1135114 25 g
AAA1135122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1135106 Supplier Thermo Scientific Chemicals Supplier No. A1135106
Only null left

CAS: 25185-95-9 | C7H5Cl2NO | 190.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25185-95-9
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.02
MDL Number MFCD00013938
InChI Key YBSXDWIAUZOFFV-ONNFQVAWSA-N
Synonym 2,6-dichlorobenzaldoxime, 2,6-dichlorobenzaldehyde oxime, 2,6-dichlorobenzaloxime, 2,6-dichloro benzyloxime, z-2,6-dichlorobenzaldehyde oxime, 2,6-dichlorophenyl hydroxyimino methane, z-n-2,6-dichlorophenyl methylidene hydroxylamine, z-2,6-dichlorobenzaldehydeoxime, e-n-2,6-dichlorophenyl methylidene hydroxylamine, e-2,6-dichlorobenzaldehyde oxime
PubChem CID 9581041
IUPAC Name (NE)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine
SMILES O\N=C\C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 147°C to 151°C
Quantity 5 g
Beilstein 2046662
Formula Weight 190.03
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorobenzaldoxime
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-720-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only