missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichlorobenzaldehyde 97.0+%, TCI America™
$104.38 - $1001.00
Chemical Identifiers
| CAS | 83-38-5 |
|---|---|
| Molecular Formula | C7H4Cl2O |
| Molecular Weight (g/mol) | 175.008 |
| MDL Number | MFCD00003307 |
| InChI Key | DMIYKWPEFRFTPY-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro |
| PubChem CID | 6737 |
| IUPAC Name | 2,6-dichlorobenzaldehyde |
| SMILES | C1=CC(=C(C(=C1)Cl)C=O)Cl |
Chemical Identifiers
| 83-38-5 | |
| 175.008 | |
| DMIYKWPEFRFTPY-UHFFFAOYSA-N | |
| 6737 | |
| C1=CC(=C(C(=C1)Cl)C=O)Cl |
| C7H4Cl2O | |
| MFCD00003307 | |
| benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro | |
| 2,6-dichlorobenzaldehyde |
Specifications
| 83-38-5 | |
| White-Yellow | |
| C7H4Cl2O | |
| 25 g | |
| benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro | |
| C1=CC(=C(C(=C1)Cl)C=O)Cl | |
| 175.008 | |
| 175.01 | |
| Crystalline Powder |
| 71°C | |
| 231°C | |
| MFCD00003307 | |
| 1759 | |
| DMIYKWPEFRFTPY-UHFFFAOYSA-N | |
| 2,6-dichlorobenzaldehyde | |
| 6737 | |
| ≥97.0% (GC) | |
| 2,6-Dichlorobenzaldehyde |
Safety and Handling
EINECSNumber : (3)-1189
TSCA : Yes