Learn More
2,6-Dichlorobenzaldehyde, 97+%
CAS: 83-38-5 | C7H4Cl2O | 175.008 g/mol
$60.19 - $757.32
Chemical Identifiers
| CAS | 83-38-5 |
|---|---|
| Molecular Formula | C7H4Cl2O |
| Molecular Weight (g/mol) | 175.008 |
| MDL Number | MFCD00003307 |
| InChI Key | DMIYKWPEFRFTPY-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro |
| PubChem CID | 6737 |
| IUPAC Name | 2,6-dichlorobenzaldehyde |
| SMILES | C1=CC(=C(C(=C1)Cl)C=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1329514
|
Thermo Scientific Chemicals
A1329514 |
25 g |
Each for $60.19
|
|
|||||
|
AAA1329522
|
Thermo Scientific Chemicals
A1329522 |
100 g |
Each for $165.95
|
|
|||||
|
AAA1329536
|
Thermo Scientific Chemicals
A1329536 |
500 g |
Each for $757.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 83-38-5 | |
| 175.008 | |
| DMIYKWPEFRFTPY-UHFFFAOYSA-N | |
| 6737 | |
| C1=CC(=C(C(=C1)Cl)C=O)Cl |
| C7H4Cl2O | |
| MFCD00003307 | |
| benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro | |
| 2,6-dichlorobenzaldehyde |
Specifications
| 83-38-5 | |
| 124°C (10 mmHg) | |
| C7H4Cl2O | |
| 25 g | |
| 386477 | |
| benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro | |
| C1=CC(=C(C(=C1)Cl)C=O)Cl | |
| 175.008 | |
| 175.01 | |
| 2,6-Dichlorobenzaldehyde |
| 69°C to 73°C | |
| 135°C (275°F) | |
| MFCD00003307 | |
| UN3261 | |
| Air Sensitive | |
| DMIYKWPEFRFTPY-UHFFFAOYSA-N | |
| 2,6-dichlorobenzaldehyde | |
| 6737 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 201-472-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only