missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroaniline 99.0+%, TCI America™

Catalog No. p-7126144
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032325G 25 g
D0323100G 100 g
D0323500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D032325G Supplier TCI America Supplier No. D032325G
Only null left

Chemical Identifiers

CAS 608-31-1
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007675
InChI Key JDMFXJULNGEPOI-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline
PubChem CID 11846
ChEBI CHEBI:46630
IUPAC Name 2,6-dichloroaniline
SMILES NC1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 228°C
Quantity 25 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloroaniline
EINECSNumber (3)-0261
TSCA Yes