missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroaniline, 98%

Catalog No. AAA1232022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1232022 100 g
AAA1232036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1232022 Supplier Thermo Scientific Chemicals Supplier No. A1232022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 608-31-1 | C6H5Cl2N | 162.01 g/mol

2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloroaniline is used in the pharmaceutical industry for the synthesis of antibacterial agents of lomefloxacin, Lee uric acid and clonidine.

Solubility
Soluble in water (1.6 g/L at 20°C), dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 608-31-1
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007675
InChI Key JDMFXJULNGEPOI-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dichloro, 2,6-dichlorobenzenamine, 2,6 dichloraniline, 2,6-dichloro aniline, 2,6-dichloranilin, 2,6-dichloro-phenylamine, 2,6-dichlorophenylamine, 2,5-dichlorobenzoic acid pestanal, clonidine impurity c, 2,6-dichloro-aniline
PubChem CID 11846
ChEBI CHEBI:46630
IUPAC Name 2,6-dichloroaniline
SMILES NC1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 37°C to 40°C
Density 1.28
Boiling Point 97°C (5 mmHg)
Flash Point 118°C (244°F)
Quantity 100 g
UN Number UN3442
Beilstein 775084
Solubility Information Soluble in water (1.6g/L at 20°C),dimethyl sulfoxide and methanol.
Formula Weight 162.02
Percent Purity 98%
Chemical Name or Material 2,6-Dichloroaniline
Show More Show Less
Hazard Category H301+H311+H331-H373-H500
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
EINECSNumber 210-160-5
RTECSNumber CX9862655
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only