missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-iodopyridine 98.0+%, TCI America™

Catalog No. D47745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47745G 5 g
D47741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47745G Supplier TCI America Supplier No. D47745G

Chemical Identifiers

CAS 98027-84-0
Molecular Formula C5H2Cl2IN
Molecular Weight (g/mol) 273.882
MDL Number MFCD07368400
InChI Key NGSKFMPSBUAUNE-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-iodo-pyridine, 2 6-dichloro-4-iodopyridine, pyridine, 2,6-dichloro-4-iodo, pubchem17648, 2,6 dichloro-4-iodo pyridine, 2,6-dichloro-4-iodopyridine,, 4-iodo-2,6-dichloropyridine, 2,6-dichloro-4-iodopyridine
PubChem CID 11737393
IUPAC Name 2,6-dichloro-4-iodopyridine
SMILES C1=C(C=C(N=C1Cl)Cl)I

Specifications

Melting Point 163°C
Quantity 5 g
Formula Weight 273.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-iodopyridine