missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-3-nitropyridine 98.0+%, TCI America™
$55.92 - $165.90
Chemical Identifiers
| CAS | 16013-85-7 |
|---|---|
| Molecular Formula | C5H2Cl2N2O2 |
| Molecular Weight (g/mol) | 192.983 |
| MDL Number | MFCD00006234 |
| InChI Key | SHCWQWRTKPNTEM-UHFFFAOYSA-N |
| Synonym | 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n |
| PubChem CID | 85239 |
| IUPAC Name | 2,6-dichloro-3-nitropyridine |
| SMILES | C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl |
Chemical Identifiers
| 16013-85-7 | |
| 192.983 | |
| SHCWQWRTKPNTEM-UHFFFAOYSA-N | |
| 85239 | |
| C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl |
| C5H2Cl2N2O2 | |
| MFCD00006234 | |
| 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n | |
| 2,6-dichloro-3-nitropyridine |
Specifications
| 16013-85-7 | |
| C5H2Cl2N2O2 | |
| 5 g | |
| SHCWQWRTKPNTEM-UHFFFAOYSA-N | |
| 2,6-dichloro-3-nitropyridine | |
| 85239 | |
| ≥98.0% (GC) | |
| 2,6-Dichloro-3-nitropyridine |
| 65°C | |
| MFCD00006234 | |
| 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n | |
| C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl | |
| 192.983 | |
| 192.98 | |
| Crystalline Powder |
Safety and Handling
TSCA : No