missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-3-nitropyridine, 97%
CAS: 16013-85-7 | C5H2Cl2N2O2 | 192.983 g/mol
$107.16 - $1,373.41
Chemical Identifiers
| CAS | 16013-85-7 |
|---|---|
| Molecular Formula | C5H2Cl2N2O2 |
| Molecular Weight (g/mol) | 192.983 |
| MDL Number | MFCD00006234 |
| InChI Key | SHCWQWRTKPNTEM-UHFFFAOYSA-N |
| Synonym | 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n |
| PubChem CID | 85239 |
| IUPAC Name | 2,6-dichloro-3-nitropyridine |
| SMILES | C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2125509
|
Thermo Scientific Chemicals
B2125509 |
10 g |
N/A
|
|
|||||
|
AAB2125518
|
Thermo Scientific Chemicals
B2125518 |
50 g |
N/A
|
|
|||||
|
AAB2125530
|
Thermo Scientific Chemicals
B2125530 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16013-85-7 | |
| 192.983 | |
| SHCWQWRTKPNTEM-UHFFFAOYSA-N | |
| 85239 | |
| C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl |
| C5H2Cl2N2O2 | |
| MFCD00006234 | |
| 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n | |
| 2,6-dichloro-3-nitropyridine |
Specifications
| 16013-85-7 | |
| >110°C (230°F) | |
| MFCD00006234 | |
| 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n | |
| C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl | |
| 192.983 | |
| 192.99 | |
| 2,6-Dichloro-3-nitropyridine |
| 62°C to 65°C | |
| C5H2Cl2N2O2 | |
| 10 g | |
| SHCWQWRTKPNTEM-UHFFFAOYSA-N | |
| 2,6-dichloro-3-nitropyridine | |
| 85239 | |
| 97% |
Safety and Handling
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 240-151-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only