missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitropyridine, 97%

Catalog No. AAB2125509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2125509 10 g
AAB2125518 50 g
AAB2125530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2125509 Supplier Thermo Scientific Chemicals Supplier No. B2125509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16013-85-7 | C5H2Cl2N2O2 | 192.983 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16013-85-7
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.983
MDL Number MFCD00006234
InChI Key SHCWQWRTKPNTEM-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n
PubChem CID 85239
IUPAC Name 2,6-dichloro-3-nitropyridine
SMILES C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 62°C to 65°C
Flash Point >110°C (230°F)
Quantity 10 g
Formula Weight 192.99
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-nitropyridine
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 240-151-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only