missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog No. p-7044395
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0618606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0618606 Supplier Thermo Scientific Chemicals Supplier No. L0618606

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5866-98-8
Molecular Formula C7H2Cl2N2O2
Molecular Weight (g/mol) 217.005
MDL Number MFCD00051513
InChI Key NSKVWZIEYFSHIM-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitrobenzenecarbonitrile, benzonitrile, 2,6-dichloro-3-nitro, acmc-1ala2, dichloronitrobenzenecarbonitrile, 2,6-dichloro-3-nitorbenzonitirle, 2,6-bis chloranyl-3-nitro-benzenecarbonitrile
PubChem CID 4461932
IUPAC Name 2,6-dichloro-3-nitrobenzonitrile
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])Cl)C#N)Cl

Specifications

Melting Point 106°C to 109°C
Quantity 5 g
UN Number UN3439
Beilstein 2113045
Formula Weight 217.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-3-nitrobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only