missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog No. p-7053674
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6127103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6127103 Supplier Thermo Scientific Chemicals Supplier No. H6127103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58584-94-4 | Molecular Formula: C6H5Cl2N | Molecular Weight: 162.01 g/mol

2,6-Dichloro-3-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-3-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 58584-94-4
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00137847
InChI Key KNIKCOPEHZTQJV-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-methyl-pyridine, pyridine, 2,6-dichloro-3-methyl, pubchem24157, 2,6-dichloro-3-picoline, acmc-1b02p, ksc608k6n, pyridine,2,6-dichloro-3-methyl, 2,6-bis chloranyl-3-methyl-pyridine, 2,6-dichloro-5-methylpyridine
PubChem CID 817088
IUPAC Name 2,6-dichloro-3-methylpyridine
SMILES CC1=C(Cl)N=C(Cl)C=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 162.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,6-Dichloro-3-methylpyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only