missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-3-cyanopyridine 98.0+%, TCI America™
$41.02 - $115.57
Chemical Identifiers
| CAS | 40381-90-6 |
|---|---|
| Molecular Formula | C6H2Cl2N2 |
| Molecular Weight (g/mol) | 172.996 |
| MDL Number | MFCD03411341 |
| InChI Key | LFCADBSUDWERJT-UHFFFAOYSA-N |
| Synonym | 2,6-dichloronicotinonitrile, 2,6-dichloro-3-cyanopyridine, 3-pyridinecarbonitrile, 2,6-dichloro, 2,6-dichloro-nicotinonitrile, 2,6-dichloro-3-pyridinecarbonitrile, 2,6-dichloropyridine-3-carbonitrile, pubchem14370, acmc-20a0s7, ksc495i4d, 3-cyano-2,6-dichloropyridine |
| PubChem CID | 12605538 |
| IUPAC Name | 2,6-dichloropyridine-3-carbonitrile |
| SMILES | C1=CC(=NC(=C1C#N)Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4650200MG
|
TCI America
D4650200MG |
200 mg |
Each for $41.02
|
|
|||||
|
D46501G
|
TCI America
D46501G |
1 g |
Each for $115.57
|
|
|||||
Chemical Identifiers
| 40381-90-6 | |
| 172.996 | |
| LFCADBSUDWERJT-UHFFFAOYSA-N | |
| 12605538 | |
| C1=CC(=NC(=C1C#N)Cl)Cl |
| C6H2Cl2N2 | |
| MFCD03411341 | |
| 2,6-dichloronicotinonitrile, 2,6-dichloro-3-cyanopyridine, 3-pyridinecarbonitrile, 2,6-dichloro, 2,6-dichloro-nicotinonitrile, 2,6-dichloro-3-pyridinecarbonitrile, 2,6-dichloropyridine-3-carbonitrile, pubchem14370, acmc-20a0s7, ksc495i4d, 3-cyano-2,6-dichloropyridine | |
| 2,6-dichloropyridine-3-carbonitrile |
Specifications
| 40381-90-6 | |
| White | |
| MFCD03411341 | |
| 3439 | |
| LFCADBSUDWERJT-UHFFFAOYSA-N | |
| 2,6-dichloropyridine-3-carbonitrile | |
| 12605538 | |
| ≥98.0% (GC) | |
| 2,6-Dichloro-3-cyanopyridine |
| 124°C | |
| C6H2Cl2N2 | |
| 200 mg | |
| 2,6-dichloronicotinonitrile, 2,6-dichloro-3-cyanopyridine, 3-pyridinecarbonitrile, 2,6-dichloro, 2,6-dichloro-nicotinonitrile, 2,6-dichloro-3-pyridinecarbonitrile, 2,6-dichloropyridine-3-carbonitrile, pubchem14370, acmc-20a0s7, ksc495i4d, 3-cyano-2,6-dichloropyridine | |
| C1=CC(=NC(=C1C#N)Cl)Cl | |
| 172.996 | |
| 173.00 | |
| Crystalline Powder |
Safety and Handling
TSCA : No