missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromoquinone-4-chloroimide 98.0+%, TCI America™

Catalog No. D02095G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D02095G Supplier TCI America Supplier No. D02095G

Chemical Identifiers

CAS 537-45-1
Molecular Formula C6H2Br2ClNO
Molecular Weight (g/mol) 299.346
MDL Number MFCD00013792
InChI Key JYWKEVKEKOTYEX-UHFFFAOYSA-N
Synonym 2,6-dibromoquinone-4-chloroimide, 2,6-dibromo-4-chloroimino cyclohexa-2,5-dienone, bqc reagent, 2,6-dibromoquinone-4-chlorimide, 2,6-dibromoquinone chlorimide, 2,6-dibromoquinone chloroimide, 2,6-dibromoquinone chloroimine, n-chloro-2,6-dibromoquinoneimine, 2,5-cyclohexadien-1-one, 2,6-dibromo-4-chloroimino, 2,6-dibromo-p-benzoquinone-4-chlorimine
PubChem CID 10835
IUPAC Name 2,6-dibromo-4-chloroiminocyclohexa-2,5-dien-1-one
SMILES C1=C(C(=O)C(=CC1=NCl)Br)Br

Specifications

Melting Point 83°C
Quantity 5 g
Formula Weight 299.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromoquinone-4-chloroimide
RTECSNumber GU5435000
TSCA Yes
Recommended Storage Refrigerator