missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyridine 98.0+%, TCI America™

Catalog No. D155525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D155525G 25 g
D1555250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D155525G Supplier TCI America Supplier No. D155525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-05-1 | C5H3Br2N | 236.894 g/mol

Chemical Identifiers

CAS 626-05-1
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00006223
InChI Key FEYDZHNIIMENOB-UHFFFAOYSA-N
Synonym pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
PubChem CID 12274
IUPAC Name 2,6-dibromopyridine
SMILES C1=CC(=NC(=C1)Br)Br

Specifications

Melting Point 119°C
Color White-Yellow
Boiling Point 249°C
Quantity 25 g
UN Number 2811
Formula Weight 236.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromopyridine
RTECSNumber US7883000
TSCA No