Learn More
2,6-Dibromopyridine, 98%
CAS: 626-05-1 | C5H3Br2N | 236.89 g/mol
Supplier: Thermo Scientific Chemicals 112911000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2, 6-Dibromopyridine | |
| 97.5 | |
| 118.0°C to 119.0°C | |
| 213°C | |
| 97.5% min. (GC) | |
| C5H3Br2N | |
| 100 g | |
| pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v | |
| FEYDZHNIIMENOB-UHFFFAOYSA-N | |
| 2,6-dibromopyridine | |
| 12274 | |
| 98% |
| 626-05-1 | |
| 100.0 | |
| Gray to White | |
| Authentic | |
| Glass bottle | |
| (C5H3N)Br2 | |
| 20, V,5, 435 | |
| Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,dioxane,chloroform,,carbon tetrachloride,heptane,aq. naoh,aq. h2so4 | |
| C1=CC(=NC(=C1)Br)Br | |
| 236.89 | |
| 236.89 | |
| Crystalline Powder |
Chemical Identifiers
| 626-05-1 | |
| 236.89 | |
| 12274 | |
| C1=CC(=NC(=C1)Br)Br |
| C5H3Br2N | |
| FEYDZHNIIMENOB-UHFFFAOYSA-N | |
| 2,6-dibromopyridine |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume
GHS Signal Word: Danger
EINECSNumber : 210-926-9
RUO – Research Use Only