missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyridine, 98%

CAS: 626-05-1 | C5H3Br2N | 236.894 g/mol

Supplier:  Thermo Scientific Chemicals A1539722

 View more versions of this product

Catalog No. AAA1539722


Only null left
Explore available promotions

Description

Description

2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.

Solubility
Soluble in hot methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool place.
Specifications

Specifications

2,6-Dibromopyridine
117°C to 120°C
213°C (415°F)
C5H3Br2N
100 g
108922
Soluble in hot methanol. Insoluble in water.
C1=CC(=NC(=C1)Br)Br
236.894
236.9
626-05-1
255°C
Recognizable
MFCD00006223
UN2811
pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
FEYDZHNIIMENOB-UHFFFAOYSA-N
2,6-dibromopyridine
12274
98%

Chemical Identifiers

626-05-1
236.894
FEYDZHNIIMENOB-UHFFFAOYSA-N
12274
C1=CC(=NC(=C1)Br)Br
C5H3Br2N
MFCD00006223
pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
2,6-dibromopyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H312-H332-H315-H319
Fatal if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H300-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 210-926-9

RTECSNumber : US7883000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only