missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromopyrazine 97.0+%, TCI America™
$70.84 - $270.29
Chemical Identifiers
| CAS | 23229-25-6 |
|---|---|
| Molecular Formula | C4H2Br2N2 |
| Molecular Weight (g/mol) | 237.88 |
| MDL Number | MFCD09834804 |
| InChI Key | JXKQTRCEKQCAGH-UHFFFAOYSA-N |
| Synonym | pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f |
| PubChem CID | 20361700 |
| IUPAC Name | 2,6-dibromopyrazine |
| SMILES | BrC1=CN=CC(Br)=N1 |
Chemical Identifiers
| 23229-25-6 | |
| 237.88 | |
| JXKQTRCEKQCAGH-UHFFFAOYSA-N | |
| 20361700 | |
| BrC1=CN=CC(Br)=N1 |
| C4H2Br2N2 | |
| MFCD09834804 | |
| pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f | |
| 2,6-dibromopyrazine |
Specifications
| 23229-25-6 | |
| White-Yellow | |
| C4H2Br2N2 | |
| 1 g | |
| JXKQTRCEKQCAGH-UHFFFAOYSA-N | |
| 2,6-dibromopyrazine | |
| 20361700 | |
| ≥97.0% (GC) | |
| 2,6-Dibromopyrazine |
| 53°C | |
| 65°C | |
| MFCD09834804 | |
| pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f | |
| BrC1=CN=CC(Br)=N1 | |
| 237.88 | |
| 237.88 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator