missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyrazine 97.0+%, TCI America™

Catalog No. D45441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45441G 1 g
D45445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45441G Supplier TCI America Supplier No. D45441G

Chemical Identifiers

CAS 23229-25-6
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.88
MDL Number MFCD09834804
InChI Key JXKQTRCEKQCAGH-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f
PubChem CID 20361700
IUPAC Name 2,6-dibromopyrazine
SMILES BrC1=CN=CC(Br)=N1

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 65°C
Quantity 1 g
Formula Weight 237.88
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromopyrazine
TSCA No
Recommended Storage Refrigerator