Learn More
2,6-Dibromopyrazine, 95%
CAS: 23229-25-6 | C4H2Br2N2 | 237.88 g/mol
$148.45 - $549.01
Chemical Identifiers
| CAS | 23229-25-6 |
|---|---|
| Molecular Formula | C4H2Br2N2 |
| Molecular Weight (g/mol) | 237.88 |
| MDL Number | MFCD09834804 |
| InChI Key | JXKQTRCEKQCAGH-UHFFFAOYSA-N |
| Synonym | pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f |
| PubChem CID | 20361700 |
| IUPAC Name | 2,6-dibromopyrazine |
| SMILES | BrC1=CN=CC(Br)=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6146803
|
Thermo Scientific Chemicals
H6146803 |
1 g |
Each for $148.45
|
|
|||||
|
AAH6146806
|
Thermo Scientific Chemicals
H6146806 |
5 g |
Each for $549.01
|
|
|||||
Description
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dibromopyrazine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 23229-25-6 | |
| 237.88 | |
| JXKQTRCEKQCAGH-UHFFFAOYSA-N | |
| 20361700 | |
| BrC1=CN=CC(Br)=N1 |
| C4H2Br2N2 | |
| MFCD09834804 | |
| pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f | |
| 2,6-dibromopyrazine |
Specifications
| 23229-25-6 | |
| C4H2Br2N2 | |
| 1 g | |
| Slightly soluble in water. | |
| BrC1=CN=CC(Br)=N1 | |
| 237.88 | |
| 237.88 | |
| Crystalline Solid |
| White to Brown | |
| MFCD09834804 | |
| pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f | |
| JXKQTRCEKQCAGH-UHFFFAOYSA-N | |
| 2,6-dibromopyrazine | |
| 20361700 | |
| 95% | |
| 2,6-Dibromopyrazine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only