missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyrazine, 95%

Catalog No. AAH6146803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6146803 1 g
AAH6146806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6146803 Supplier Thermo Scientific Chemicals Supplier No. H6146803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23229-25-6 | C4H2Br2N2 | 237.88 g/mol

2,6-Dibromopyrazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 23229-25-6
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.88
MDL Number MFCD09834804
InChI Key JXKQTRCEKQCAGH-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f
PubChem CID 20361700
IUPAC Name 2,6-dibromopyrazine
SMILES BrC1=CN=CC(Br)=N1

Specifications

Color White to Brown
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 237.88
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 2,6-Dibromopyrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only